Methyl 2-bromo-3,6-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

33709-1G 33709-25G 33709-5G
76940-572EA 82.76 USD
76940-572 76940-574 76940-576
Methyl 2-bromo-3,6-difluorobenzoate ≥97%
Methyl 2-bromo-3,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28061358
CAS Number: 1805523-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR