Methyl 4-chloro-2,6-dibromobenzoate ≥97%

Supplier: AOBChem USA

Warning

33616-1G 33616-500MG 33616-5G
76940-532EA 1040.75 USD
76940-532 76940-534 76940-536
Methyl 4-chloro-2,6-dibromobenzoate ≥97%
Methyl 4-chloro-2,6-dibromobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Br₂ClO₂
MW: 328.385 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561305
CAS Number: 1332271-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR