Methyl 3-chloro-5-fluoro-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

33537-1G 33537-500MG 33537-5G
76940-506EA 759.61 USD
76940-506 76940-508 76940-510
Methyl 3-chloro-5-fluoro-2-methoxybenzoate ≥95%
Methyl 3-chloro-5-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382552
CAS Number: 2105066-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR