Ethyl 2-bromo-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

33531-1G 33531-25G 33531-500MG 33531-5G
76940-492EA 343.74 USD
76940-492 76940-494 76940-496 76940-498
Ethyl 2-bromo-6-methoxybenzoate ≥97%
Ethyl 2-bromo-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD14698172
CAS Number: 1214387-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR