Methyl 2-fluoro-4-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

33503-1G 33503-25G 33503-5G
76940-478EA 62.6 USD
76940-478 76940-480 76940-482
Methyl 2-fluoro-4-(trifluoromethyl)benzoate ≥97%
Methyl 2-fluoro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06203705
CAS Number: 220162-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR