Ethyl 5-chloro-2-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

33315-1G 33315-25G 33315-500MG 33315-5G
76940-424EA 351.16 USD
76940-424 76940-426 76940-428 76940-430
Ethyl 5-chloro-2-ethoxybenzoate ≥97%
Ethyl 5-chloro-2-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994148
CAS Number: 73119-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR