Methyl 4-formyl-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

33012-1G 33012-25G 33012-500MG 33012-5G
76940-392EA 393.6 USD
76940-392 76940-394 76940-396 76940-398
Methyl 4-formyl-3-methoxybenzoate ≥97%
Methyl 4-formyl-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17169951
CAS Number: 74733-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR