2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

32988-1G 32988-25G 32988-5G
76940-370EA 62.6 USD
76940-370 76940-372 76940-374
2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97% research grade
2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
Formula: C₁₄H₁₈BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD12923177
CAS Number: 757982-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR