6-Fluorobenzo[b]thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

32883-1G 32883-25G 32883-5G
76940-360EA 115.64 USD
76940-360 76940-362 76940-364
6-Fluorobenzo[b]thiophene-2-carboxylic acid ≥97%

Precaución: for Research Use Only

Formula: C₉H₅FO₂S
Storage Temperature: Ambient
MDL Number: MFCD06655861
CAS Number: 142329-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR