Ethyl 5-fluoro-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

32867-1G 32867-25G 32867-500MG 32867-5G
76940-346EA 274.78 USD
76940-346 76940-348 76940-350 76940-352
Ethyl 5-fluoro-2-methoxybenzoate ≥95%
Ethyl 5-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06204449
CAS Number: 773135-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR