Methyl 2-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32671-1G 32671-25G 32671-500MG 32671-5G
76940-298EA 266.29 USD
76940-298 76940-300 76940-302 76940-304
Methyl 2-methoxy-3-methylbenzoate ≥97%
Methyl 2-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025375
CAS Number: 52239-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR