6-Chloro-2-iodo-3-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-iodopyridin-3-ol

32615-25G 32615-5G
76940-276EA 315.09 USD
76940-276 76940-278
6-Chloro-2-iodo-3-hydroxypyridine ≥97%
6-Chloro-2-iodo-3-hydroxypyridine
Formula: C₅H₃ClIO
MW: 255.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07636749
CAS Number: 188057-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR