Methyl 2,6-dibromo-4-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

32540-1G 32540-25G 32540-5G
76940-264EA 96.55 USD
76940-264 76940-266 76940-268
Methyl 2,6-dibromo-4-fluorobenzoate ≥97%
Methyl 2,6-dibromo-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Br₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD28784668
CAS Number: 1806294-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR