Ethyl 5-bromo-2,4-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32400-1G 32400-25G 32400-500MG 32400-5G
76940-212EA 290.69 USD
76940-212 76940-214 76940-216 76940-218
Ethyl 5-bromo-2,4-dimethoxybenzoate ≥97%
Ethyl 5-bromo-2,4-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₄
Storage Temperature: Refrigerator
MDL Number: MFCD06204646
CAS Number: 773136-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR