Ethyl 3-bromo-2,6-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32298-1G 32298-25G 32298-500MG 32298-5G
76940-178EA 291.75 USD
76940-178 76940-180 76940-182 76940-184
Ethyl 3-bromo-2,6-dimethoxybenzoate ≥97%
Ethyl 3-bromo-2,6-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₄
MW: 289.123 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11870186
CAS Number: 1352206-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR