Methyl 2-chloro-4-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32252-1G 32252-25G 32252-500MG 32252-5G
76940-162EA 159.14 USD
76940-162 76940-164 76940-166 76940-168
Methyl 2-chloro-4-formylbenzoate ≥97%
Methyl 2-chloro-4-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD24685927
CAS Number: 908248-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR