8-(5-Bromopyridin-2-yl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

32150-1G 32150-25G 32150-5G
76940-094EA 79.57 USD
76940-094 76940-096 76940-098
8-(5-Bromopyridin-2-yl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(5-Bromopyridin-2-yl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28964012
CAS Number: 708274-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR