6-Chloro-5-methoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-5-methoxypyridine-3-carboxylic acid, 3-Carboxy-6-chloro-5-methoxypyridine

32023-1G 32023-25G 32023-500MG 32023-5G
76940-058EA 153.84 USD
76940-058 76940-060 76940-062 76940-064
6-Chloro-5-methoxynicotinic acid ≥97%
6-Chloro-5-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD12828186
CAS Number: 915107-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR