6-Bromo-2,3-dichlorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31808-1G 31808-25G 31808-500MG 31808-5G
76940-020EA 350.1 USD
76940-020 76940-022 76940-024 76940-026
6-Bromo-2,3-dichlorobenzyl alcohol ≥97%
6-Bromo-2,3-dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD28737840
CAS Number: 1807009-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR