Methyl 2-bromo-4,6-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

31618-1G 31618-500MG 31618-5G
76939-986EA 474.23 USD
76939-986 76939-988 76939-990
Methyl 2-bromo-4,6-difluorobenzoate ≥97%
Methyl 2-bromo-4,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18642939
CAS Number: 1379336-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR