1-Chloro-3,5-difluoro-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2,4-difluoroiodobenzene

31582-1G 31582-25G 31582-500MG 31582-5G
76939-972EA 296 USD
76939-972 76939-974 76939-976 76939-978
1-Chloro-3,5-difluoro-2-iodobenzene ≥95%
1-Chloro-3,5-difluoro-2-iodobenzene
Formula: C₆H₂ClF₂I
MW: 274.434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817642
CAS Number: 1242339-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR