Ethyl 3-fluoro-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

31454-1G 31454-25G 31454-500MG 31454-5G
76939-960EA 142.17 USD
76939-960 76939-962 76939-964 76939-966
Ethyl 3-fluoro-5-(trifluoromethyl)benzoate ≥97%
Ethyl 3-fluoro-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06204413
CAS Number: 250682-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR