Methyl-3-bromo-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-bromo-5-methoxybenzoate

31436-1G 31436-25G 31436-500MG 31436-5G
76939-944EA 245.07 USD
76939-944 76939-946 76939-948 76939-950
Methyl-3-bromo-5-methoxybenzoate ≥97%
Methyl 3-bromo-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD11042779
CAS Number: 56709-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR