6-Bromo-3-methoxypicolinic acid ≥97%

Supplier: AOBChem USA

Warning

31342-1G 31342-25G 31342-500MG 31342-5G
76939-920EA 122.01 USD
76939-920 76939-922 76939-924 76939-926
6-Bromo-3-methoxypicolinic acid ≥97%
6-Bromo-3-methoxypicolinic acid
Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD11846420
CAS Number: 1256810-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR