Methyl 2,6-dimethoxypyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

31325-1G 31325-25G 31325-500MG 31325-5G
76939-904EA 231.28 USD
76939-904 76939-906 76939-908 76939-910
Methyl 2,6-dimethoxypyridine-4-carboxylate ≥97%
Methyl 2,6-dimethoxypyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MW: 197.188 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04111112
CAS Number: 52606-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR