Ethyl 5-bromo-4-chloronicotinate ≥95%

Supplier: AOBChem USA

Warning

31168-1G 31168-500MG
76939-880EA 1679.41 USD
76939-880 76939-882
Ethyl 5-bromo-4-chloronicotinate ≥95%
Ethyl 5-bromo-4-chloronicotinate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.504 g/mol
Storage Temperature: Ambient
CAS Number: 1805519-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR