Ethyl 4-bromothiophene-3-carboxylate ≥95% research grade

Supplier: AOBChem USA

Warning

31157-1G 31157-25G 31157-500MG 31157-5G
76939-864EA 124.13 USD
76939-864 76939-866 76939-868 76939-870
Ethyl 4-bromothiophene-3-carboxylate ≥95% research grade
Ethyl 4-bromothiophene-3-carboxylate
Formula: C₇H₇BrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD14584478
CAS Number: 224449-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR