Methyl 4-methoxy-1-naphthoate ≥97%

Supplier: AOBChem USA

Warning

31143-1G 31143-25G 31143-500MG 31143-5G
76939-854EA 142.17 USD
76939-854 76939-856 76939-858 76939-860
Methyl 4-methoxy-1-naphthoate ≥97%
Methyl 4-methoxy-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₃
MW: 216.233 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203661
CAS Number: 13041-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR