6-Bromo-2-fluoro-3-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31104-25G 31104-5G
76939-846EA 519.85 USD
76939-846 76939-848
6-Bromo-2-fluoro-3-methylphenylboronic acid pinacol ester ≥97%
6-Bromo-2-fluoro-3-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MW: 314.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088259
CAS Number: 2121514-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR