Methyl 3,5-difluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

30929-1G 30929-25G 30929-500MG 30929-5G
76939-810EA 142.17 USD
76939-810 76939-812 76939-814 76939-816
Methyl 3,5-difluoro-4-methylbenzoate ≥97%
Methyl 3,5-difluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28053776
CAS Number: 1806321-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR