Methyl 5-bromo-2,4-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

30904-1G 30904-25G 30904-5G
76939-796EA 91.24 USD
76939-796 76939-798 76939-800
Methyl 5-bromo-2,4-difluorobenzoate ≥97%
Methyl 5-bromo-2,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD16038827
CAS Number: 351325-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR