6-Chloro-3-fluoro-2-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chlorol-3-fluoro-2-iodopyridine

30542-1G 30542-25G 30542-500MG 30542-5G
76939-718EA 320.4 USD
76939-718 76939-720 76939-722 76939-724
6-Chloro-3-fluoro-2-iodopyridine ≥97%
6-Chloro-3-fluoro-2-iodopyridine
Formula: C₅H₂ClFIN
MW: 257.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257562
CAS Number: 1211590-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR