Methyl 2-ethoxypyridine-3-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
30529-1G
30529-25G
30529-5G
76939-706EA
84.88
USD
76939-706
76939-708
76939-710
Methyl 2-ethoxypyridine-3-carboxylate ≥97%
Methyl 2-ethoxypyridine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₃ MW: 181.189 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20527397 CAS Number: 74357-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...