Methyl 2-bromo-5-chloronicotinate ≥97%

Supplier: AOBChem USA

Warning

30424-1G 30424-25G 30424-500MG 30424-5G
76939-676EA 260.99 USD
76939-676 76939-678 76939-680 76939-682
Methyl 2-bromo-5-chloronicotinate ≥97%
Methyl 2-bromo-5-chloronicotinate
Formula: C₇H₅BrClNO₂
MW: 250.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698100
CAS Number: 1214324-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR