Methyl 4-bromo-2,3-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

30353-1G 30353-25G 30353-500MG 30353-5G
76939-664EA 155.96 USD
76939-664 76939-666 76939-668 76939-670
Methyl 4-bromo-2,3-dimethylbenzoate ≥97%
Methyl 4-bromo-2,3-dimethylbenzoate
Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23703480
CAS Number: 6021-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR