Ethyl 2-chloro-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

30305-1G 30305-25G 30305-5G
76939-650EA 96.55 USD
76939-650 76939-652 76939-654
Ethyl 2-chloro-5-fluorobenzoate ≥97%
Ethyl 2-chloro-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06205162
CAS Number: 57381-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR