N,N-Dimethyl-3-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N,N-Dimethyl-3-trifluoromethylaniline

30175-1G 30175-25G 30175-5G
76939-636EA 81.69 USD
76939-636 76939-638 76939-640
N,N-Dimethyl-3-(trifluoromethyl)aniline ≥97%
N,N-Dimethyl-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632487
CAS Number: 329-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR