6-Phenylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

30167-1G 30167-25G 30167-5G
76939-624EA 96.55 USD
76939-624 76939-626 76939-628
6-Phenylnicotinic acid ≥97%
6-Phenylnicotinic acid
Formula: C₁₂H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD03086128
CAS Number: 29051-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR