Ethyl 3-bromo-5-ethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

30133-1G 30133-25G 30133-5G
76939-596EA 62.6 USD
76939-596 76939-598 76939-600
Ethyl 3-bromo-5-ethoxybenzoate ≥97%
Ethyl 3-bromo-5-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₃
MW: 273.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26403777
CAS Number: 1229442-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR