Methyl 2-chloro-6-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

29970-1G 29970-25G 29970-500MG 29970-5G
76939-532EA 290.69 USD
76939-532 76939-534 76939-536 76939-538
Methyl 2-chloro-6-fluoro-3-methoxybenzoate ≥97%
Methyl 2-chloro-6-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566180
CAS Number: 1379356-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR