tert-Butyl (5-bromopyridin-3-yl)carbamate ≥95% research grade

Supplier: AOBChem USA

Warning

29755-25G 29755-5G
76939-506EA 198.39 USD
76939-506 76939-508
tert-Butyl (5-bromopyridin-3-yl)carbamate ≥95% research grade
tert-Butyl (5-bromopyridin-3-yl)carbamate
Formula: C₁₀H₁₃BrN₂O₂
MW: 273.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367900
CAS Number: 361550-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR