Cyclohexanone-p-toluenesulfonylhydrazone ≥95%

Supplier: AOBChem USA

Warning

29625-25G 29625-5G
76939-482EA 198.39 USD
76939-482 76939-484
Cyclohexanone-p-toluenesulfonylhydrazone ≥95%
Cyclohexanone-p-toluenesulfonylhydrazone
Formula: C₁₃H₁₈N₂O₂S
Storage Temperature: Ambient
CAS Number: 4545-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR