N-(2-Bromo-4,5-difluorophenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

29507-1G 29507-25G 29507-5G
76939-474EA 62.6 USD
76939-474 76939-476 76939-478
N-(2-Bromo-4,5-difluorophenyl)acetamide ≥97%
N-(2-Bromo-4,5-difluorophenyl)acetamide
Formula: C₈H₆BrF₂NO
MW: 250.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705814
CAS Number: 64695-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR