6-Bromo-5-methoxypyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

29385-1G 29385-25G 29385-500MG 29385-5G
76939-448EA 260.99 USD
76939-448 76939-450 76939-452 76939-454
6-Bromo-5-methoxypyridine-3-carboxylic acid ≥95%
6-Bromo-5-methoxypyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257274
CAS Number: 1256810-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR