6-Bromo-2,3-dihydro-1H-inden-1-ol ≥95%

Supplier: AOBChem USA

Warning

28900-25G 28900-5G
76939-398EA 587.74 USD
76939-398 76939-400
6-Bromo-2,3-dihydro-1H-inden-1-ol ≥95%
6-Bromo-2,3-dihydro-1H-inden-1-ol
Formula: C₉H₉BrO
Storage Temperature: Refrigerator
MDL Number: MFCD02916451
CAS Number: 75476-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR