Methyl 3-bromo-5-chloro-2-fluorobenzoate ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
28789-1G
28789-25G
28789-500MG
28789-5G
76939-384EA
231.28
USD
76939-384
76939-386
76939-388
76939-390
Methyl 3-bromo-5-chloro-2-fluorobenzoate ≥97% research grade
Methyl 3-bromo-5-chloro-2-fluorobenzoate
Formula:
C₈H₅BrClFO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD24107111 CAS Number: 1353575-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...