Methyl 3,6-difluoro-2-hydroxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

28742-1G 28742-25G 28742-5G
76939-370EA 106.09 USD
76939-370 76939-372 76939-374
Methyl 3,6-difluoro-2-hydroxybenzoate ≥97% research grade
Methyl 3,6-difluoro-2-hydroxybenzoate
Formula: C₈H₆F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD10567189
CAS Number: 1214324-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR