Ethyl 3-bromoisonicotinate ≥95% research grade

Supplier: AOBChem USA

Warning

28705-1G 28705-25G 28705-5G
76939-356EA 62.6 USD
76939-356 76939-358 76939-360
Ethyl 3-bromoisonicotinate ≥95% research grade
Ethyl 3-bromoisonicotinate
Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11226640
CAS Number: 13959-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR