5-Iodo-2-methylbenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

28550-25G 28550-5G
76939-350EA 488.02 USD
76939-350 76939-352
5-Iodo-2-methylbenzotrifluoride ≥97%
5-Iodo-2-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆F₃I
Storage Temperature: Ambient
MDL Number: MFCD12026291
CAS Number: 930599-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR