5-Propyl-thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

28379-1G 28379-25G 28379-500MG 28379-5G
76939-328EA 185.66 USD
76939-328 76939-330 76939-332 76939-334
5-Propyl-thiophene-2-carboxylic acid ≥97%
5-Propyl-thiophene-2-carboxylic acid
Formula: C₈H₁₀O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD04970762
CAS Number: 63068-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR